The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020598.2 | NP_065623.2 | 1 - 1 | 553 | 78 |
GenBank mRNA | BC139721.1 | - | 1 - 1 | 553 | 78 |