The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021433.3 | NP_067408.1 | 1 - 2 | 228 | 63 |
GenBank mRNA | AF237814.1 | - | 1 - 2 | 38 | 63 |
AK009690.1 | - | 1 - 2 | 183 | 63 | |
AK019106.1 | - | 1 - 2 | 190 | 63 | |
AK031673.1 | - | 1 - 2 | 207 | 63 | |
AK044905.1 | - | 1 - 2 | 189 | 63 | |
AK049124.1 | - | 1 - 2 | 188 | 63 | |
AK151549.1 | - | 1 - 2 | 182 | 63 | |
AK159392.1 | - | 1 - 2 | 200 | 63 | |
BC029205.1 | - | 1 - 2 | 145 | 63 | |
BQ749205.1 | - | 1 - 2 | 201 | 63 | |
CJ088919.1 | - | 1 - 2 | 228 | 63 |