Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a7
Entrez Gene ID
63993
Gene Name
solute carrier family 5 (choline transporter), member 7
Gene Aliases
CHT1
Chromosome Location
Chr.17: 54273590 - 54299041 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022025.4 NP_071308.2 5 - 6 1099 70
GenBank mRNA AF276872.2 - 4 - 5 590 70
AJ401467.1 - 4 - 5 590 70
AK034415.1 - 5 - 6 983 70
AK053063.1 - 5 - 6 1101 70
BC065089.1 - 4 - 5 616 70

View Full Product Details