The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022025.4 | NP_071308.2 | 8 - 9 | 1622 | 72 |
GenBank mRNA | AF276872.2 | - | 7 - 8 | 1113 | 72 |
AJ401467.1 | - | 7 - 8 | 1113 | 72 | |
AK034415.1 | - | 8 - 9 | 1506 | 72 | |
AK053063.1 | - | 8 - 9 | 1624 | 72 | |
AK164116.1 | - | 1 - 2 | 530 | 72 | |
BC065089.1 | - | 7 - 8 | 1139 | 72 |