Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thop1
Entrez Gene ID
50492
Gene Name
thimet oligopeptidase 1
Gene Aliases
AI131655, AI327041, EP24.15
Chromosome Location
Chr.10: 81070083 - 81082360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022653.4 NP_073144.3 6 - 7 922 69
GenBank mRNA AF314187.1 - 6 - 7 880 69
AK028609.1 - 6 - 7 856 69
CN723672.1 - 4 - 5 467 69

View Full Product Details