The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199113.1 | NP_001186042.1 | 7 - 8 | 830 | 76 |
NM_001199114.1 | NP_001186043.1 | 6 - 7 | 669 | 76 | |
NM_001199115.1 | NP_001186044.1 | 6 - 7 | 775 | 76 | |
NM_001199116.1 | NP_001186045.1 | 6 - 7 | 735 | 76 | |
NM_022880.3 | NP_075018.1 | 5 - 6 | 776 | 76 | |
GenBank mRNA | AF131212.1 | - | 5 - 6 | 777 | 76 |
AF305501.1 | - | 6 - 7 | 740 | 76 | |
AK004756.1 | - | 6 - 7 | 721 | 76 | |
AK050089.1 | - | 6 - 7 | 760 | 76 | |
AK146265.1 | - | 6 - 7 | 670 | 76 | |
AK149483.1 | - | 6 - 7 | 691 | 76 | |
AK170463.1 | - | 6 - 7 | 707 | 76 | |
AK181560.1 | - | 3 - 4 | 315 | 76 | |
AK193260.1 | - | 2 - 3 | 210 | 76 | |
AK214211.1 | - | 2 - 3 | 210 | 76 | |
BC004828.1 | - | 7 - 8 | 793 | 76 | |
BC006812.1 | - | 5 - 6 | 707 | 76 | |
EU180577.1 | - | 5 - 6 | 645 | 76 |