The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204906.1 | NP_001191835.1 | 2 - 3 | 109 | 74 |
NM_001204907.1 | NP_001191836.1 | 2 - 3 | 109 | 74 | |
NM_023042.3 | NP_075529.2 | 2 - 3 | 109 | 74 | |
GenBank mRNA | AB017104.1 | - | 1 - 2 | 70 | 74 |
AB017105.1 | - | 1 - 2 | 62 | 74 | |
AK137589.1 | - | 2 - 3 | 109 | 74 | |
AK138325.1 | - | 2 - 3 | 65 | 74 | |
AK138721.1 | - | 2 - 3 | 102 | 74 | |
AK164344.1 | - | 2 - 3 | 79 | 74 |