Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Palm
Entrez Gene ID
18483
Gene Name
paralemmin
Gene Aliases
-
Chromosome Location
Chr.10: 79793351 - 79820896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161747.1 NP_001155219.1 2 - 3 221 85
NM_023128.4 NP_075617.3 2 - 3 221 85
GenBank mRNA AK037044.1 - 2 - 3 78 85
AK082163.1 - 2 - 3 223 85
AK133733.1 - 2 - 3 127 85
AK162411.1 - 2 - 3 214 85
BC015297.1 - 2 - 3 205 85
Y14771.1 - 2 - 3 184 85

View Full Product Details