Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Qsox1
Entrez Gene ID
104009
Gene Name
quiescin Q6 sulfhydryl oxidase 1
Gene Aliases
1300003H02Rik, QSOX, Qscn6, SOx
Chromosome Location
Chr.1: 155778155 - 155812899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024945.1 NP_001020116.1 4 - 5 627 105
NM_023268.2 NP_075757.1 4 - 5 627 105
GenBank mRNA AB044284.1 - 4 - 5 535 105
AK004880.1 - 4 - 5 573 105
AK083938.1 - 4 - 5 578 105
AK166839.1 - 4 - 5 573 105
AK169877.1 - 4 - 5 580 105
AK169920.1 - 4 - 5 576 105
AK170449.1 - 4 - 5 540 105
BC034131.1 - 1 - 2 119 105
BC076590.1 - 4 - 5 581 105

View Full Product Details