Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upk3a
Entrez Gene ID
22270
Gene Name
uroplakin 3A
Gene Aliases
1110017C07Rik, UP3a, UPIII, Upk3
Chromosome Location
Chr.15: 85017141 - 85022560 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023478.2 NP_075967.2 5 - 6 787 60
GenBank mRNA AF222750.2 - 5 - 6 708 60
AK003737.1 - 3 - 4 298 60
BC125336.1 - 5 - 6 787 60
BC125338.1 - 5 - 6 787 60
BY703997.1 - 3 - 4 298 60
DQ387454.1 - 5 - 6 737 60

View Full Product Details