The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252591.2 | NP_001239520.1 | 1 - 2 | 336 | 66 |
NR_045569.1 | 1 - 2 | 336 | 66 | ||
NR_103722.1 | 1 - 2 | 336 | 66 | ||
GenBank mRNA | AA434782.1 | - | 1 - 2 | 191 | 66 |
AF364070.1 | - | 1 - 2 | 175 | 66 | |
AI035961.1 | - | 1 - 2 | 150 | 66 | |
AJ245772.1 | - | 1 - 2 | 191 | 66 | |
AK003105.1 | - | 1 - 2 | 183 | 66 | |
AK010172.1 | - | 1 - 2 | 183 | 66 | |
AY026524.1 | - | 1 - 2 | 182 | 66 | |
BC051197.1 | - | 1 - 2 | 135 | 66 | |
BC061051.1 | - | 1 - 2 | 97 | 66 | |
W18646.1 | - | 1 - 2 | 49 | 66 |