Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pccb
Entrez Gene ID
66904
Gene Name
propionyl Coenzyme A carboxylase, beta polypeptide
Gene Aliases
1300012P06Rik, AI314687, R74805
Chromosome Location
Chr.9: 100982032 - 101034940 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025835.2 NP_080111.1 14 - 15 1521 66
GenBank mRNA AF327060.1 - 14 - 15 1509 66
AK004468.1 - 2 - 3 134 66
AK004976.1 - 14 - 15 1522 66
AK050366.1 - 14 - 15 1527 66
AK134437.1 - 13 - 14 1424 66
BC026531.1 - 14 - 15 1497 66
BC055946.1 - 14 - 15 1528 66

View Full Product Details