Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan4
Entrez Gene ID
64540
Gene Name
tetraspanin 4
Gene Aliases
AI325509, AI746565, D130042I01Rik, NAG-2, Tm4sf7, Tspan-4
Chromosome Location
Chr.7: 141475236 - 141493427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252588.1 NP_001239517.1 4 - 5 443 65
NM_053082.3 NP_444312.1 4 - 5 606 65
GenBank mRNA AK002709.1 - 4 - 5 366 65
AK156959.1 - 4 - 5 562 65
AK172481.1 - 4 - 5 424 65
AK184168.1 - - 169 65
AK189309.1 - 1 - 2 81 65
AK211721.1 - 3 - 4 337 65
BC003482.1 - 4 - 5 386 65
BC083070.1 - 4 - 5 602 65
CB954815.1 - 4 - 5 609 65
CT010285.1 - 3 - 4 327 65

View Full Product Details