Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Recql4
Entrez Gene ID
79456
Gene Name
RecQ protein-like 4
Gene Aliases
RecQ4
Chromosome Location
Chr.15: 76703553 - 76710576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058214.3 NP_478121.2 16 - 17 2863 74
GenBank mRNA AB175741.1 - 16 - 17 2780 74
BC064680.1 - 16 - 17 2837 74
BC131656.1 - 16 - 17 2776 74
BC131657.1 - 14 - 15 3113 74

View Full Product Details