The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133231.2 | NP_573494.1 | 1 - 2 | 594 | 73 |
GenBank mRNA | AF335512.1 | - | 1 - 2 | 574 | 73 |
AK077640.1 | - | 1 - 2 | 592 | 73 | |
AK144633.1 | - | 1 - 2 | 581 | 73 | |
BC019935.1 | - | 1 - 2 | 557 | 73 | |
BC047123.1 | - | 1 - 2 | 557 | 73 |