The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286007.1 | NP_001272936.1 | 8 - 9 | 1576 | 91 |
NM_001286009.1 | NP_001272938.1 | 8 - 9 | 1576 | 91 | |
NM_001286011.1 | NP_001272940.1 | 7 - 8 | 1343 | 91 | |
NM_133755.2 | NP_598516.2 | 8 - 9 | 1503 | 91 | |
GenBank mRNA | AK033157.1 | - | 8 - 9 | 1293 | 91 |
AK045084.1 | - | 8 - 9 | 1292 | 91 | |
AK146771.1 | - | 8 - 9 | 1498 | 91 | |
AK153511.1 | - | 8 - 9 | 1424 | 91 | |
AK158060.1 | - | 8 - 9 | 1292 | 91 | |
AK167924.1 | - | 8 - 9 | 1398 | 91 | |
AK186148.1 | - | 1 - 2 | 112 | 91 | |
BC012519.1 | - | 8 - 9 | 1431 | 91 | |
BC025564.1 | - | 7 - 8 | 1265 | 91 | |
BC025582.1 | - | 8 - 9 | 1392 | 91 | |
BC027034.1 | - | 8 - 9 | 1487 | 91 |