Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcnxl3
Entrez Gene ID
104401
Gene Name
pecanex-like 3 (Drosophila)
Gene Aliases
mKIAA4073
Chromosome Location
Chr.19: 5664635 - 5688908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144868.3 NP_659117.2 28 - 29 5462 78
GenBank mRNA AF237953.1 - 26 - 27 3479 78
AK144114.1 - 4 - 5 722 78
AK220471.1 - 6 - 7 1360 78
BC038056.1 - 23 - 24 3570 78
BC054751.1 - 28 - 29 4741 78
BC055289.1 - 29 - 30 5265 78
BC096538.1 - 28 - 29 5462 78

View Full Product Details