Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp3
Entrez Gene ID
235441
Gene Name
ubiquitin specific peptidase 3
Gene Aliases
AA409661, BC017156
Chromosome Location
Chr.9: 66514639 - 66593115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK031141.1 - 5 - 6 560 101
AK158516.1 - 3 - 4 388 101
AK161676.1 - 5 - 6 617 101
BC017156.1 - 5 - 6 681 101

View Full Product Details