Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pi4k2a
Entrez Gene ID
84095
Gene Name
phosphatidylinositol 4-kinase type 2 alpha
Gene Aliases
Pi4k2
Chromosome Location
Chr.19: 42090146 - 42122218 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145501.2 NP_663476.1 6 - 7 1152 56
GenBank mRNA AK030725.1 - 6 - 7 1117 56
AK185438.1 - 2 - 3 173 56
BC110363.1 - 6 - 7 1114 56

View Full Product Details