The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145587.2 | NP_663562.1 | 2 - 3 | 713 | 120 |
GenBank mRNA | BC024114.1 | - | 2 - 3 | 454 | 120 |
BC025837.1 | - | 2 - 3 | 550 | 120 | |
BC031759.1 | - | 2 - 3 | 713 | 120 |