Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plcd4
Entrez Gene ID
18802
Gene Name
phospholipase C, delta 4
Gene Aliases
4921507K24Rik
Chromosome Location
Chr.1: 74541972 - 74565977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148937.2 NP_683739.2 11 - 12 1892 77
GenBank mRNA AY033991.1 - 11 - 12 1724 77

View Full Product Details