The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148945.2 | NP_683747.1 | 15 - 16 | 1688 | 99 |
GenBank mRNA | AK154778.1 | - | 15 - 16 | 1600 | 99 |
AY083469.1 | - | 15 - 16 | 1350 | 99 | |
BC150156.1 | - | 15 - 16 | 1353 | 99 | |
BC150478.1 | - | 15 - 16 | 1353 | 99 |