The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007528.3 | NP_031554.1 | 5 - 6 | 977 | 70 |
GenBank mRNA | AB011665.1 | - | 4 - 5 | 900 | 70 |
AK156733.1 | - | 5 - 6 | 939 | 70 | |
BC090998.1 | - | 5 - 6 | 937 | 70 |