The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009212.2 | NP_033238.2 | 1 - 2 | 312 | 101 |
GenBank mRNA | AK142456.1 | - | 1 - 2 | 116 | 101 |
AK153763.1 | - | 1 - 2 | 273 | 101 | |
AK154805.1 | - | 1 - 2 | 309 | 101 | |
AK155899.1 | - | 1 - 2 | 309 | 101 | |
BC052025.1 | - | 1 - 2 | 282 | 101 | |
BC060085.1 | - | 1 - 2 | 282 | 101 | |
BC083142.1 | - | 1 - 2 | 117 | 101 | |
BC133681.1 | - | 1 - 2 | 120 | 101 | |
L10075.1 | - | 1 - 2 | 208 | 101 |