Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Avil
Entrez Gene ID
11567
Gene Name
advillin
Gene Aliases
Advil, DOC6
Chromosome Location
Chr.10: 127000075 - 127020994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009635.3 NP_033765.2 2 - 3 210 90
GenBank mRNA AF041448.1 - 2 - 3 161 90
AF059486.1 - 2 - 3 180 90
AK154851.1 - 2 - 3 181 90
AK155900.1 - 2 - 3 181 90
AK170878.1 - 2 - 3 181 90
AK171319.1 - 2 - 3 182 90
AK171324.1 - 2 - 3 181 90
BC119222.1 - 2 - 3 115 90
BC120545.1 - 2 - 3 115 90

View Full Product Details