Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Angpt1
Entrez Gene ID
11600
Gene Name
angiopoietin 1
Gene Aliases
1110046O21Rik, Ang-1, Ang1
Chromosome Location
Chr.15: 42424667 - 42676977 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286062.1 NP_001272991.1 3 - 4 1096 118
NM_009640.4 NP_033770.2 3 - 4 1096 118
GenBank mRNA AK012027.1 - 3 - 4 1032 118
AK088439.1 - 3 - 4 1097 118
AK172868.1 - 3 - 4 916 118
BC067410.1 - 3 - 4 914 118
U83509.1 - 3 - 4 886 118

View Full Product Details