The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206367.1 | NP_001193296.1 | 11 - 12 | 1531 | 60 |
NM_001206368.1 | NP_001193297.1 | 10 - 11 | 1447 | 60 | |
NM_001206369.1 | NP_001193298.1 | 10 - 11 | 1492 | 60 | |
NM_146120.4 | NP_666232.2 | 10 - 11 | 1554 | 60 | |
GenBank mRNA | AK076156.1 | - | 10 - 11 | 1494 | 60 |
AK089934.1 | - | 11 - 12 | 1486 | 60 | |
AK143664.1 | - | 10 - 11 | 1521 | 60 | |
AK146969.1 | - | 11 - 12 | 1448 | 60 | |
AK148957.1 | - | 11 - 12 | 1511 | 60 | |
AK150359.1 | - | 11 - 12 | 1454 | 60 | |
AK151684.1 | - | 11 - 12 | 1473 | 60 | |
AK153029.1 | - | 11 - 12 | 1451 | 60 | |
AK166831.1 | - | 10 - 11 | 1467 | 60 | |
AK166932.1 | - | 11 - 12 | 1445 | 60 | |
AK167918.1 | - | 11 - 12 | 1461 | 60 | |
AK168461.1 | - | 11 - 12 | 1464 | 60 | |
AK168532.1 | - | 11 - 12 | 1465 | 60 | |
AK168707.1 | - | 10 - 11 | 1400 | 60 | |
AK168725.1 | - | 11 - 12 | 1448 | 60 | |
AK168829.1 | - | 10 - 11 | 1499 | 60 | |
AK169041.1 | - | 11 - 12 | 1451 | 60 | |
AK169670.1 | - | 11 - 12 | 1476 | 60 | |
BC023143.1 | - | 10 - 11 | 1492 | 60 | |
BC060377.1 | - | 11 - 12 | 1479 | 60 | |
J04953.1 | - | 9 - 10 | 60 |