The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010722.5 | NP_034852.2 | 5 - 6 | 879 | 58 |
GenBank mRNA | AK132671.1 | - | 2 - 3 | 587 | 58 |
BC042430.1 | - | 5 - 6 | 884 | 58 | |
BC051985.1 | - | 5 - 6 | 855 | 58 | |
D13455.1 | - | 2 - 3 | 566 | 58 | |
X54098.1 | - | 5 - 6 | 838 | 58 |