Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sptb
Entrez Gene ID
20741
Gene Name
spectrin beta, erythrocytic
Gene Aliases
AI842465, D330027P03Rik, Gm1301, Spnb-1, Spnb1, ja, jaundiced, mKIAA4219
Chromosome Location
Chr.12: 76580488 - 76710547 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013675.3 NP_038703.3 2 - 3 819 95
GenBank mRNA AK147299.1 - 2 - 3 820 95
AK147684.1 - 2 - 3 412 95
AK147704.1 - 2 - 3 412 95
BC117711.2 - 1 - 2 145 95
BC129806.1 - 1 - 2 152 95
BC150783.1 - 1 - 2 185 95
S66283.1 - 2 - 3 295 95

View Full Product Details