The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013786.2 | NP_038814.1 | 5 - 6 | 934 | 70 |
GenBank mRNA | AK008212.1 | - | 5 - 6 | 993 | 70 |
BC021836.1 | - | 5 - 6 | 934 | 70 |