Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zpbp
Entrez Gene ID
53604
Gene Name
zona pellucida binding protein
Gene Aliases
4930486K01Rik, Iam38, Sp38
Chromosome Location
Chr.11: 11279624 - 11463630 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185153.1 NP_001172082.1 4 - 5 537 154
NM_015785.2 NP_056600.1 4 - 5 708 154
GenBank mRNA BC139450.1 - 4 - 5 677 154
BC139453.1 - 4 - 5 677 154
BC145628.1 - 4 - 5 506 154
D17569.1 - 4 - 5 697 154

View Full Product Details