The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146049.1 | NP_001139521.1 | 5 - 6 | 767 | 86 |
NM_001146050.1 | NP_001139522.1 | 5 - 6 | 601 | 86 | |
NM_001146052.1 | NP_001139524.1 | 4 - 5 | 591 | 86 | |
NM_001146053.1 | NP_001139525.1 | 5 - 6 | 545 | 86 | |
NM_016865.3 | NP_058561.2 | 5 - 6 | 711 | 86 | |
GenBank mRNA | AF061972.1 | - | 4 - 5 | 695 | 86 |
AK008630.1 | - | 4 - 5 | 586 | 86 | |
AK144837.1 | - | 5 - 6 | 745 | 86 | |
AK146400.1 | - | 4 - 5 | 406 | 86 | |
AK151254.1 | - | 5 - 6 | 632 | 86 | |
AK152418.1 | - | 5 - 6 | 632 | 86 | |
BB575839.2 | - | 5 - 6 | 545 | 86 | |
BC004083.1 | - | 5 - 6 | 664 | 86 | |
BC017372.1 | - | 4 - 5 | 557 | 86 | |
CB318388.1 | - | 4 - 5 | 594 | 86 | |
CB593552.1 | - | 5 - 6 | 593 | 86 |