Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hpx
Entrez Gene ID
15458
Gene Name
hemopexin
Gene Aliases
Hpxn, hx
Chromosome Location
Chr.7: 105591611 - 105600116 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017371.2 NP_059067.2 3 - 4 239 65
GenBank mRNA AK145928.1 - 3 - 4 237 65
BC011246.1 - 3 - 4 219 65
BC019901.1 - 3 - 4 221 65
BY088352.1 - 3 - 4 240 65
U89889.1 - 3 - 4 195 65

View Full Product Details