The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023603.3 | NP_076092.1 | 3 - 4 | 1410 | 68 |
NR_045010.1 | 3 - 4 | 1410 | 68 | ||
GenBank mRNA | AF110499.1 | - | 1 - 2 | 192 | 68 |
AF272847.1 | - | 3 - 4 | 703 | 68 | |
AK157783.1 | - | 4 - 5 | 1047 | 68 | |
AK179158.1 | - | 1 - 2 | 98 | 68 | |
AK180570.1 | - | 1 - 2 | 69 | ||
AK211570.1 | - | 2 - 3 | 350 | 68 | |
AK214936.1 | - | 1 - 2 | 52 | 68 | |
AY034062.1 | - | 3 - 4 | 1325 | 68 | |
BC037486.1 | - | 1 - 2 | 123 | 68 | |
BC080848.1 | - | 3 - 4 | 1376 | 68 | |
BC089305.1 | - | 3 - 4 | 1376 | 68 | |
CA319647.1 | - | 1 - 2 | 41 | 68 |