The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025399.3 | NP_079675.1 | 1 - 2 | 128 | 112 |
GenBank mRNA | AK003991.1 | - | 1 - 2 | 135 | 112 |
AK013174.1 | - | 1 - 2 | 84 | 112 | |
BC025444.1 | - | 1 - 2 | 130 | 112 |