The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026257.3 | NP_080533.1 | 7 - 8 | 526 | 57 |
NR_045571.1 | 7 - 8 | 526 | 57 | ||
GenBank mRNA | AK015720.1 | - | 7 - 8 | 510 | 57 |
AK048494.1 | - | 7 - 8 | 528 | 57 | |
BC019461.1 | - | 7 - 8 | 503 | 57 |