The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026474.5 | NP_080750.1 | 12 - 13 | 1053 | 68 |
GenBank mRNA | AK007881.1 | - | 12 - 13 | 988 | 68 |
AK010830.1 | - | 12 - 13 | 970 | 68 | |
AK020433.1 | - | 12 - 13 | 972 | 68 | |
AK045153.1 | - | 12 - 13 | 992 | 68 | |
AK082328.1 | - | 12 - 13 | 1055 | 68 | |
AK141416.1 | - | - | 989 | 68 | |
AK148838.1 | - | 12 - 13 | 967 | 68 | |
BC009167.1 | - | 12 - 13 | 970 | 68 | |
BC038677.1 | - | - | 451 | 68 |