Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rufy2
Entrez Gene ID
70432
Gene Name
RUN and FYVE domain-containing 2
Gene Aliases
2610111M19Rik, AI844453, Denn, LZ-FYVE, ZFYVE13
Chromosome Location
Chr.10: 62980223 - 63018742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027425.3 NP_081701.2 13 - 14 1427 78
GenBank mRNA AK173195.1 - 13 - 14 1388 78
BC137736.1 - 13 - 14 1376 78
BC137737.1 - 13 - 14 1376 78

View Full Product Details