The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027493.3 | NP_081769.2 | 12 - 13 | 1860 | 77 |
GenBank mRNA | AK017815.1 | - | 12 - 13 | 1811 | 77 |
BC027281.1 | - | 12 - 13 | 1766 | 77 |