The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029556.3 | NP_083832.2 | 7 - 8 | 955 | 74 |
GenBank mRNA | AF428254.1 | - | 7 - 8 | 928 | 74 |
AK002741.1 | - | 7 - 8 | 1083 | 74 | |
AK009345.1 | - | 7 - 8 | 943 | 74 | |
BC023398.1 | - | 7 - 8 | 945 | 74 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AW123793.1 | - | Chr. - - [-] on Build | 74 |