The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133946.2 | NP_598707.2 | 4 - 5 | 2238 | 135 |
GenBank mRNA | AK039531.1 | - | 1 - 2 | 1522 | 135 |
AY355343.1 | - | 1 - 2 | 1176 | 135 | |
BC013519.1 | - | 1 - 2 | 1145 | 135 | |
BC031139.1 | - | 1 - 2 | 1230 | 135 | |
FJ222825.1 | - | 4 - 5 | 2184 | 135 | |
JF810536.1 | - | 3 - 4 | 2623 | 135 |