Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Catsper1
Entrez Gene ID
225865
Gene Name
cation channel, sperm associated 1
Gene Aliases
Catsper, KSper
Chromosome Location
Chr.19: 5335602 - 5344281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139301.2 NP_647462.1 9 - 10 1843 82
GenBank mRNA AF407332.1 - 9 - 10 1843 82

View Full Product Details