The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146198.2 | NP_666310.1 | 4 - 5 | 483 | 83 |
GenBank mRNA | AF518726.1 | - | 5 - 6 | 576 | 83 |
AK008219.1 | - | 4 - 5 | 537 | 83 | |
BC031742.1 | - | 4 - 5 | 484 | 83 |