The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146207.2 | NP_666319.2 | 11 - 12 | 1270 | 73 |
GenBank mRNA | AK144802.1 | - | 11 - 12 | 1253 | 74 |
AK150237.1 | - | 11 - 12 | 1249 | 73 | |
AK168215.1 | - | 11 - 12 | 1252 | 73 | |
AK170722.1 | - | 11 - 12 | 1252 | 73 | |
BC010211.1 | - | 11 - 12 | 1248 | 73 | |
BC024113.1 | - | 7 - 8 | 742 | 73 | |
BC027006.1 | - | 7 - 8 | 790 | 73 |