Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spg7
Entrez Gene ID
234847
Gene Name
spastic paraplegia 7 homolog (human)
Gene Aliases
AI452278, AU015315, Cmar, PGN
Chromosome Location
Chr.8: 123065508 - 123097751 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153176.4 NP_694816.3 12 - 13 1683 90
GenBank mRNA AF512565.1 - 12 - 13 1715 90
AF547215.1 - 12 - 13 2012 90
AK145540.1 - 12 - 13 1684 90
AK171647.1 - 9 - 10 2517 90
BC024466.1 - 6 - 7 744 90
BC024986.1 - 10 - 11 1338 90
BC051051.1 - 12 - 13 1617 90
BC055488.1 - - 1528 90
BC096690.1 - 5 - 6 583 90
BC138141.1 - 12 - 13 1910 90

View Full Product Details