The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290824.1 | NP_001277753.1 | 15 - 16 | 1960 | 101 |
NM_153387.3 | NP_700436.1 | 15 - 16 | 1963 | 101 | |
NR_077245.2 | 15 - 16 | 1960 | 101 | ||
NR_110985.1 | 15 - 16 | 1963 | 101 | ||
GenBank mRNA | AK034694.1 | - | 6 - 7 | 1491 | 101 |
AK038921.1 | - | 15 - 16 | 1919 | 101 | |
AK045305.1 | - | 15 - 16 | 1856 | 101 | |
AK046132.1 | - | 15 - 16 | 1893 | 101 | |
AK050250.1 | - | 15 - 16 | 1961 | 101 | |
AK152984.1 | - | 15 - 16 | 1919 | 101 | |
AK153171.1 | - | 15 - 16 | 1919 | 101 | |
AK161774.1 | - | 6 - 7 | 759 | 101 | |
BC029106.1 | - | 15 - 16 | 1927 | 101 |