Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adam32
Entrez Gene ID
353188
Gene Name
a disintegrin and metallopeptidase domain 32
Gene Aliases
-
Chromosome Location
Chr.8: 24836143 - 24948917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293693.1 NP_001280622.1 12 - 13 1248 95
NM_153397.2 NP_700446.2 12 - 13 1248 95
GenBank mRNA AF513715.1 - 12 - 13 1231 95
AK005759.1 - 12 - 13 1239 95
BC060983.1 - 12 - 13 1245 95

View Full Product Details