Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc65
Entrez Gene ID
105833
Gene Name
coiled-coil domain containing 65
Gene Aliases
4933417K04Rik, AI428139, AU020178, NYD-SP28
Chromosome Location
Chr.15: 98708227 - 98723336 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153518.1 NP_705738.1 6 - 7 1210 85
GenBank mRNA AF454432.1 - 6 - 7 1210 85
AK077174.1 - 6 - 7 1244 85
BC048073.1 - 6 - 7 1230 85

View Full Product Details