Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkag3
Entrez Gene ID
241113
Gene Name
protein kinase, AMP-activated, gamma 3 non-catatlytic subunit
Gene Aliases
AMPKg3L, AMPKg3S, Amkg3, Ampkg3
Chromosome Location
Chr.1: 74738922 - 74749590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153744.3 NP_714966.1 3 - 4 332 76
GenBank mRNA AF525500.1 - 3 - 4 239 76
AF525501.1 - 2 - 3 260 76
AK036585.1 - 3 - 4 333 76
BC116749.1 - 2 - 3 196 76
BC116777.1 - 2 - 3 196 76

View Full Product Details