Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkag3
Entrez Gene ID
241113
Gene Name
protein kinase, AMP-activated, gamma 3 non-catatlytic subunit
Gene Aliases
AMPKg3L, AMPKg3S, Amkg3, Ampkg3
Chromosome Location
Chr.1: 74738922 - 74749590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153744.3 NP_714966.1 8 - 9 977 71
GenBank mRNA AF525500.1 - 8 - 9 884 71
AF525501.1 - 7 - 8 905 71
AK036585.1 - 8 - 9 978 71
BC116749.1 - 7 - 8 841 71
BC116777.1 - 7 - 8 841 71

View Full Product Details